accessibility menu, dialog, popup

UserName

CAS RN 1006302-79-9

NClFFClOOH

CAS RN 1006302-79-9

IUPAC Name: 2-(3,5-dichloro-2,6-difluoropyridin-4-yl)acetic acid
Synonyms: (3,5-dichloro-2,6-difluoropyridin-4-yl)acetic acid
Molecular Weight (g/mol): 242
Molecular Formula: C7H3Cl2F2NO2
InChi Key: SGLXACHKHUYVJQ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(Cl)C(F)=NC(F)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results