accessibility menu, dialog, popup

UserName

CAS RN 1003575-43-6

BOOH3CCH3H3CH3CFNH2

CAS RN 1003575-43-6

IUPAC Name: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 237.08
Molecular Formula: C12H17BFNO2
InChi Key: VXNIVUKZNDRJBW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results