CAS RN 10031-87-5
CAS RN 10031-87-5
IUPAC Name:
2-ethylbutyl acetate
Synonyms:
1-butanol, 2-ethyl-, acetate
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
HQLKZWRSOHTERR-UHFFFAOYSA-N
SMILES:
CCC(CC)COC(C)=O
1
–
1
of
1
results
1
2-Ethylbutyl acetate, 97%, Thermo Scientific™
CAS: 10031-87-5 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.214 MDL Number: MFCD00009343 InChI Key: HQLKZWRSOHTERR-UHFFFAOYSA-N Synonym: acetic acid, 2-ethylbutyl ester,2-ethyl butyl acetate,beta-ethylbutyl acetate,acetic acid 2-ethylbutyl ester,1-butanol, 2-ethyl-, acetate,fema no. 2425,2-ethylbutylacetat,2-ethylbutyl=acetate,acmc-2097ot,2-ethylbutyl acetate PubChem CID: 61448 IUPAC Name: 2-ethylbutyl acetate SMILES: CCC(CC)COC(=O)C