missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZZZ3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101968
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZZZ3.
Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
Specifications
| ZZZ3 | |
| Polyclonal | |
| Unconjugated | |
| ZZZ3 | |
| 3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC component 1 homolog; ATAC1; AW556059; DKFZP564I052; R75514; RGD1565468; zinc finger ZZ-type containing 3; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3; Zzz3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108946, 26009, 310958 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6KAQ7, Q8IYH5 | |
| ZZZ3 | |
| A synthesized peptide derived from human ZZZ3(Accession Q8IYH5), corresponding to amino acid residues Y199-T249. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction