missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ZZL 7
GREENER_CHOICE

Product Code. 30000035
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 30000035

Brand: Tocris Bioscience™ 7898/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of the interaction between the 5-HT transporter (SERT) and neuronal nitric oxide synthase (nNOS)

ZZL-7 is a fast-acting antidepressant that enhances 5-HT (serotonin) signaling in forebrain circuits. ZZL-7 disrupts the interaction between the 5-HT transporter (SERT) and neuronal nitric oxide synthase (nNOS) in the dorsal raphe nucleus (DRN), which reduces extracellular 5-HT concentration and down-regulates 5-HT1A receptor auto-activity in the DRN. This reduced auto-inhibition enables increase in firing frequency of serotonergic neurons in the DRN and increases in 5-HT release in the medial hippocampus. ZZL-7 can readily cross the blood-brain barrier.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Acetyl-L-alanyl-L-valine methyl ester
CAS 99141-91-0
Quantity 10 mg
Target SLC6A4/5-HTTLPR/Serotonin transporter
Molecular Formula C11H20N2O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.