missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZZEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556860
This item is not returnable.
View return policy
Description
Immunogen sequence: LNDRSLLPAL SCVQTALLHL LDMGWEPNDL AFFVDIQLPD LLMKMSQENI SVHDSVISQW SEEDELADAK QNSEWMDECQ DGMF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.
ZZEF1 gene ontology annotations related to this gene include calcium ion binding; zinc ion binding.
Specifications
| ZZEF1 | |
| Polyclonal | |
| Unconjugated | |
| ZZEF1 | |
| FLJ10821; KIAA0399; zinc finger ZZ-type and EF-hand domain containing 1; zinc finger ZZ-type and EF-hand domain-containing protein 1; zinc finger, ZZ-type with EF hand domain 1; zinc finger, ZZ-type with EF-hand domain 1; ZZEF1; ZZZ4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23140 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43149 | |
| ZZEF1 | |
| Recombinant protein corresponding to Human ZZEF1. Recombinant protein control fragment (Product #RP-95572). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction