missing translation for 'onlineSavingsMsg'
Learn More

Zyxin Antibody (2C10-4A7), Novus Biologicals™

Product Code. 18362579 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18362579 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18362579 Supplier Novus Biologicals Supplier No. H00007791M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Zyxin Monoclonal antibody specifically detects Zyxin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zyxin
Applications Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, KnockDown
Classification Monoclonal
Clone 2C10-4A7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH08743
Gene Alias CTCL tumor antigen se14-3, CTCL-associated antigen se14-3, cutaneous T-cell lymphoma associated antigen se14-3, Cutaneous T-cell lymphoma-associated antigen se14-3, KIAA1125, MGC31836, predicted protein of HQ2893, PRKCBP1, PRO2893, protein kinase C-binding protein 1, Rack7, RACK7protein kinase C binding protein 1, zinc finger MYND domain containing protein 8, Zinc finger MYND domain-containing protein 8, zinc finger, MYND-type containing 8
Host Species Mouse
Immunogen ZYX (AAH08743, 1 a.a. ~ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAPRPSPAISVSVSAPAFYAPQEKFGPVVAPKPKVNPFRPGDSEPPPAPGAQRAQMGRVGEIPPPPPEDFPLPPPPLAGDGDDAEGALGGAFPPPPPPIEESFPPAPLEEEIFPSPPPPPEEEGGPEAPIPPPPQPREKVSSIDLEIDSLSSLLDDMTKNDPFKARVSSGYVPPPVATPFSSKSSTKPAAGGTAPLPPWKSPSSSQPLPQVPAPAQSQTQFHVQPQPQPKPQVQLHVQSQTQPVSLANTQPRGPPASSPAPAPKFSPVTPKFTPVASKFSPGAPGGSGSQPNQKLGHPEALSAGTGSPQPPSFTYAQQREKPRVQEKQHPVPPPAQNQNQVRSPGAPGPLTLKEVEELEQLTQQLMQDMEHPQRQNVAVNELCGRCHQPLARAQPAVRALGQLFHIACFTCHQCAQQLQGQQFYSLEGAPYCEGCYTDTLEKCNTCGEPITDRMLRATGKAYHPHCFTCVVCARPLEGTSFIVDQANRPHCVPDYHKQYAPRCSVCSEPIMPEPGRDETVRVVALDKNFHMKCYECEDCGKPLSIEADDNGCFPLDGHVLCRKCHTARAQT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7791
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.