missing translation for 'onlineSavingsMsg'
Learn More

ZXH-3-26, MedChemExpress

Product Code. 16495796
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495796 100 mg 100mg
16495058 10 mg 10mg
16461297 50 mg 50mg
16437787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495796 Supplier Medchem Express Supplier No. HY122826
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ZXH-3-26 is a PROTAC connected by ligands for Cereblon and BRD4 with a DC50/5h of 5 nM. The DC50/5h refers to half-maximal degradation after 5 hours of treatment of ∼ 5 nM.

ZXH-3-26 is a PROTAC connected by ligands for Cereblon and BRD4 with a DC50/5h of 5 nM. The DC50/5h refers to half-maximal degradation after 5 hours of treatment of ∼ 5 nM.

Specifications

Chemical Name or Material ZXH-3-26
CAS 2243076-67-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C38H37ClN8O7S
Quantity 100 mg
Solubility Information DMSO : 200 mg/mL (254.69 mM; Need ultrasonic)
SMILES O=C(OC)C[C@H]1C2=NN=C(C)N2C3=C(C(C)=C(C(NCCCCCNC4=CC=CC(C(N5C(CC6)C(NC6=O)=O)=O)=C4C5=O)=O)S3)C(C7=CC=C(Cl)C=C7)=N1
Molecular Weight (g/mol) 785.27
Formula Weight 785.27
Percent Purity 98.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.