missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZXDC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543570
This item is not returnable.
View return policy
Description
Peptide sequence: NLKAHMKGHE QESLFKCEVC AERFPTHAKL SSHQRSHFEP ERPYKCDFPG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.
ZXDC belongs to the ZXD family. It contains 10 C2H2-type zinc fingers. ZXDC cooperates with CIITA to promote transcription of MHC class I and MHC class II genes.
Specifications
| ZXDC | |
| Polyclonal | |
| Unconjugated | |
| ZXDC | |
| A930012H20; B930086F11Rik; BC003332; LOW QUALITY PROTEIN: zinc finger protein ZXDC; RGD1561296; SERH2790; zinc finger protein ZXDC; ZXD family zinc finger C; ZXD family zinc finger C-like; ZXDC; ZXDL; ZXD-like zinc finger protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 79364 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q2QGD7 | |
| ZXDC | |
| Synthetic peptide directed towards the middle region of human ZXDC. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction