missing translation for 'onlineSavingsMsg'
Learn More

ZXDA Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18411028 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411028 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411028 Supplier Novus Biologicals Supplier No. NBP204129JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZXDA Polyclonal antibody specifically detects ZXDA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZXDA
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias zinc finger X-linked protein ZXDA, zinc finger, X-linked, duplicated A, ZNF896
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to this antibody and Zinc Finger, X-linked, Duplicated B using the numbering given in entry NP_009087.1 and NP_009088.1 (GeneID 7789 and 158586).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7789
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.