missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZWINT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118377
This item is not returnable.
View return policy
Description
ZWINT Polyclonal Antibody for Western Blot, IP
This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ZWINT | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| O95229 | |
| ZWINT | |
| E. coli-derived Human ZWINT fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ZWINT | |
| 2010007E07Rik; 2600001N01Rik; D10Ertd749e; human ZW10 interacting protein-1; HZwint-1; KNTC2AP; neuroprotective protein 8; scoilin; Sip30; SNAP25 interacting protein 30; SNAP25 interacting protein of 30 kDa; SNAP25-interacting protein 30; ZW10 interacting kinetochore protein; ZW10 interacting protein-1; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; ZWINT; ZWINT1; Zwint-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 11130 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction