missing translation for 'onlineSavingsMsg'
Learn More

ZWINT Antibody (1B7), Novus Biologicals™

Product Code. 18355289 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355289 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355289 Supplier Novus Biologicals Supplier No. H00011130M04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZWINT Monoclonal antibody specifically detects ZWINT in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZWINT
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1B7
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH20979
Gene Alias C20orf164, chromosome 20 open reading frame 164, dJ337O18.7, zinc finger SWIM domain-containing protein 3, zinc finger, SWIM domain containing 3, zinc finger, SWIM-type containing 3
Host Species Mouse
Immunogen ZWINT (AAH20979, 1 a.a. ~ 277 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEAAETEAEAAALEVLAEVAGILEPVGLQEEAELPAKILVEFVVDSQKKDKLLCSQLQVADFLQNILAQEDTAKGLDPLASEDTSRQKAIAAKEQWKELKATYREHVEAIKIGLTKALTQMEEAQRKRTQLREAFEQLQAKKQMAMEKRRAVQNQWQLQQEKHLQHLAEVSAEVRERKTGTQQELDGVFQKLGNLKQQAEQERDKLQRYQTFLQLLYTLQGKLLFPEAEAEAENLPDDKPQQPTRPQEQSTGDTMGRDPGVSFKAVGLQPAGDVNLP
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 11130
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.