missing translation for 'onlineSavingsMsg'
Learn More

ZW10 Antibody, Novus Biologicals™

Product Code. 18369908 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369908 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369908 Supplier Novus Biologicals Supplier No. H00009183D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZW10 Polyclonal antibody specifically detects ZW10 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZW10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. NP_004715.1
Gene Alias E3 ubiquitin-protein ligase ZSWIM2, EC 6.3.2.-, MEKK1-related protein X, MEX, MGC33890, Zinc finger SWIM domain-containing protein 2, zinc finger, SWIM domain containing 2, zinc finger, SWIM-type containing 2, ZZZ2ZZ-type zinc finger-containing protein 2
Host Species Rabbit
Immunogen ZW10 (NP_004715.1, 1 a.a. - 779 a.a.) full-length human protein. MASFVTEVLAHSGRLEKEDLGTRISRLTRRVEEIKGEVCNMISKKYSEFLPSMQSAQGLITQVDKLSEDIDLLKSRIESEVRRDLHVSTGEFTDLKQQLERDSVVLSLLKQLQEFSTAIEEYNCALTEKKYVTGAQRLEEAQKCLKLLKSRKCFDLKILKSLSMELTIQKQNILYHLGEEWQKLIVWKFPPSKDTSSLESYLQTELHLYTEQSHKEEKTPMPPISSVLLAFSVLGELHSKLKSFGQMLLKYILRPLASCPSLHAVIESQPNIVIIRFESIMTNLEYPSPSEVFTKIRLVLEVLQKQLLDLPLDTDLENEKTSTVPLAEMLGDMIWEDLSECLIKNCLVYSIPTNSSKLQQYEEIIQSTEEFENALKEMRFLKGDTTDLLKYARNINSHFANKKCQDVIVAARNLMTSEIHNTVKIIPDSKINVPELPTPDEDNKLEVQKVSNTQYHEVMNLEPENTLDQHSFSLPTCRISESVKKLMELAYQTLLEATTSSDQCAVQLFYSVRNIFHLFHDVVPTYHKENLQKLPQLAAIHHNNCMYIAHHLLTLGHQFRLRLAPILCDGTATFVDLVPGFRRLGTECFLAQMRAQKGELLERLSSARNFSNMDDEENYSAASKAVRQVLHQLKRLGIVWQDVLPVNIYCKAMGTLLNTAISEVIGKITALEDISTEDGDRLYSLCKTVMDEGPQVFAPLSEESKNKKYQEEVPVYVPKWMPFKELMMMLQASLQEIGDRWADGKGPLAAAFSSSEVKALIRALFQNTERRAAALAKIK
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 9183
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.