missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZUFSP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567602
This item is not returnable.
View return policy
Description
Specficity: The antibody detects endogenous levels of total ZUFSP protein.
Deubiquitinase with endodeubiquitinase activity that specifically interacts with and cleaves 'Lys-63'-linked long polyubiquitin chains. Shows only weak activity against 'Lys-11' and 'Lys-48'-linked chains. Plays an important role in genome stability pathways, functioning to prevent spontaneous DNA damage and also promote cellular survival in response to exogenous DNA damage. Modulates the ubiquitination status of replication protein A (RPA) complex proteins in response to replication stress.
Specifications
| ZUFSP | |
| Polyclonal | |
| Unconjugated | |
| ZUFSP | |
| 2700019D07Rik; C6orf113; dJ412I7.3; DUB; Lys-63-specific deubiquitinase ZUFSP; zinc finger with UFM1 specific peptidase domain; zinc finger with UFM1-specific peptidase domain; zinc finger with UFM1-specific peptidase domain protein; Zinc finger-containing ubiquitin peptidase 1; Zufsp; Zup1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221302 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q96AP4 | |
| ZUFSP | |
| Synthesized peptide derived from human ZUFSP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction