missing translation for 'onlineSavingsMsg'
Learn More

Zuclopenthixol, MedChemExpress

Product Code. 16414197
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16414197 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16414197 Supplier Medchem Express Supplier No. HYA0163
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Zuclopenthixol is a thioxanthene derivative which acts as a mixed dopamine D1/D2 receptor antagonist.

Zuclopenthixol is a thioxanthene derivative which acts as a mixed dopamine D1/D2 receptor antagonist.

Specifications

Chemical Name or Material Zuclopenthixol
Physical Form Solid
CAS 53772-83-1
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H25ClN2OS
For Use With (Application) Neuroscience-Neuromodulation
Synonym (Z)-Clopenthixol
Solubility Information DMSO : ≥ 150 mg/mL (374.10 mM)
SMILES OCCN1CCN(CC/C=C2C3=C(SC4=C\2C=CC=C4)C=CC(Cl)=C3)CC1
Molecular Weight (g/mol) 400.96
Formula Weight 400.96
Percent Purity 98.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.