missing translation for 'onlineSavingsMsg'
Learn More

Zuclopenthixol, Lipomed

Product Code. 30457815
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457815 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457815 Supplier Lipomed Supplier No. LPMZUC192310

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Zuclopenthixol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zuclopenthixol
CAS 53772-83-1
Analyte or Component Names Zuclopenthixol
Shipping Condition Room Temperature
Molecular Formula C22 H25 Cl N2 O S
Synonym 1-Piperazineethanol, 4-[(3 Z)-3-(2-chloro-9 H-thioxanthen-9-ylidene)propyl]-, 1-Piperazineethanol, 4-[3-(2-chloro-9 H-thioxanthen-9-ylidene)propyl]-, (Z)-, 4-[(3 Z)-3-(2-Chloro-9 H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol, 9 H-Thioxanthene, 1-piperazineethanol deriv., (Z)-Clopenthixol, Acuphase, Cisordinol, Clopixol, Clopixol depo, Zuclopenthixol, cis-(Z)-Clopenthixol, cis-Clopenthixol, alpha-Clopenthixol
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES OCCN1CCN(CC\C=C/2 \c3ccccc3Sc4ccc(Cl)cc24)CC1
IUPAC Name 2-[4-[(3 Z)-3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]ethanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 400.96
Quantity 10 mg
Formula Weight 400.1376 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.