missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zuclopenthixol-d4 Succinate Salt, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Zuclopenthixol-d4 Succinate Salt |
| CAS | 1246833-97-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C26H27D4ClN2O5S |
| Quantity | 10 mg |
| Synonym | 4-[(3Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)propyl]-1-piperazineethanol-d4 Succinate, (Z)-Clopenthixol-d4 Succinate, Cisordinol-d4, Succinate Clopixol-d4 Succinate, cis-(Z)-Clopenthixol-d4 Succinate, cis-Clopenthixol-d4 Succinate, α-Clopenthixol-d4 Succinate |
| InChI Formula | InChI=1S/C22H25ClN2OS.C4H6O4/c23-17-7-8-22-20(16-17)18(19-4-1-2-6-21(19)27-22)5-3-9-24-10-12-25(13-11-24)14-15-26;5-3(6)1-2-4(7)8/h1-2,4-8,16,26H,3,9-15H2;1-2H2,(H,5,6)(H,7,8)/i14D2,15D2; |
| SMILES | O=C(O)CCC(O)=O.OC([2H])([2H])C([2H])([2H])N(CC1)CCN1CC/C=C2C3=C(C=CC(Cl)=C3)SC4=CC=CC=C4\2 |
| IUPAC Name | butanedioic acid;2-[4-[3-(2-chlorothioxanthen-9-ylidene)propyl]piperazin-1-yl]-1,1,2,2-tetradeuterioethanol |
| Molecular Weight (g/mol) | 523.08 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction