missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572669
This item is not returnable.
View return policy
Description
CCP2 Polyclonal Antibody for Western Blot
Metallocarboxypeptidase that mediates deglutamylation of target proteins. Catalyzes the deglutamylation of polyglutamate side chains generated by post-translational polyglutamylation in proteins such as tubulins. Also removes gene-encoded polyglutamates from the carboxy-terminus of target proteins such as MYLK. Does not show detyrosinase or deglycylase activities from the carboxy-terminus of tubulin.
Specifications
| CCP2 | |
| Polyclonal | |
| Unconjugated | |
| AGBL2 | |
| 4930524K04; A430081C19Rik; Agbl2; ATP/GTP binding protein like 2; ATP/GTP binding protein-like 2; ATP/GTP-binding protein-like 2; carboxypeptidase 2, cytosolic; CBCP2; CCP2; cytoplasmic carboxypeptidase 2; cytosolic carboxypeptidase 2; testis tissue sperm-binding protein Li 96mP; testis-expressed protein 25; zebrafish testis-expressed 25; zgc:165648; zte25 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 568152 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.76 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| A6H8T7 | |
| AGBL2 | |
| KLH conjugated synthetic peptide between 92-126 amino acids of DANRE zte25. | |
| 400 μL | |
| Primary | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction