missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZSWIM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559861
This item is not returnable.
View return policy
Description
Immunogen sequence: PLDPIGCLCR ALLEACRLEE ETLTLYPDSG PEKRKVAYQH VPVPGSPGES YLVL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.
Specifications
| ZSWIM4 | |
| Polyclonal | |
| Unconjugated | |
| ZSWIM4 | |
| Zinc finger SWIM domain-containing protein 4; zinc finger SWIM-type containing 4; zinc finger, SWIM domain containing 4; zinc finger, SWIM-type containing 4; ZSWIM4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65249 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7M6 | |
| ZSWIM4 | |
| Recombinant protein corresponding to Human ZSWIM4. Recombinant protein control fragment (Product #RP-99096). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction