missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZSWIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590142
This item is not returnable.
View return policy
Description
Immunogen sequence: DMLPAQWTAG CATSLDSILG SKWSETLDKH LAVTHLTEEV GQLLQHCTKE EFERRYSTLR ELADSWIGPY EQVQL; Positive Samples: Mouse kidney.
Specifications
| ZSWIM1 | |
| Polyclonal | |
| Unconjugated | |
| ZSWIM1 | |
| 2410003H12Rik; AI850991; BB046916; C20orf162; Zinc finger SWIM domain-containing protein 1; zinc finger SWIM-type containing 1; zinc finger, SWIM domain containing 1; zinc finger, SWIM-type containing 1; Zswim1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 71971 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.17 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9CWV7 | |
| ZSWIM1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 411-485 of human ZSWIM1 (NP_5421703). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction