missing translation for 'onlineSavingsMsg'
Learn More

ZSTK474, MedChemExpress

Product Code. 16488108
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488108 10 mg 10mg
16464027 50 mg 50mg
16437876 100 mg 100mg
16421326 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488108 Supplier Medchem Express Supplier No. HY50847

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZSTK474 is an ATP-competitive pan-class I PI3K inhibitor with IC50s of16 nM, 44 nM, 4.6 nM and 49 nM for Pι3κα, PI3Kβ, PI3Kδ and PI3Kγ, respectively.

ZSTK474 is an ATP-competitive pan-class I PI3K inhibitor with IC50s of16 nM, 44 nM, 4.6 nM and 49 nM for Pι3κα, PI3Kβ, PI3Kδ and PI3Kγ, respectively.

Specifications

Chemical Name or Material ZSTK474
CAS 475110-96-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21F2N7O2
Quantity 10 mg
Solubility Information DMSO : 2 mg/mL (4.79 mM; Need ultrasonic)
SMILES FC(C1=NC2=C(N1C3=NC(N4CCOCC4)=NC(N5CCOCC5)=N3)C=CC=C2)F
Molecular Weight (g/mol) 417.41
Formula Weight 417.41
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.