missing translation for 'onlineSavingsMsg'
Learn More

ZSET1446, MedChemExpress

Product Code. 16474358
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474358 10 mg 10mg
16453046 25 mg 25mg
16451139 1 mg 1mg
16437077 5 mg 5mg
16430687 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16474358 Supplier Medchem Express Supplier No. HY11013
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ZSET1446 is a novel cognitive enhancer that significantly improves learning deficits in various types of Alzheimer disease (AD) models.

ZSET1446 is a novel cognitive enhancer that significantly improves learning deficits in various types of Alzheimer disease (AD) models.

Specifications

Chemical Name or Material ZSET1446
CAS 887603-94-3
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C15H12N2O
Quantity 10 mg
Synonym ST-101
Solubility Information DMSO : ≥ 34 mg/mL (143.90 mM)
SMILES O=C1N=C2C=CC=CN2C13CC4=C(C=CC=C4)C3
Molecular Weight (g/mol) 236.27
Formula Weight 236.27
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.