missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZSCAN5 Antibody (PCRP-ZSCAN5A-2H4) [mFluor Violet 610 SE], Novus Biologicals™
Shop All Bio Techne ProductsDescription
ZSCAN5 Monoclonal antibody specifically detects ZSCAN5 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry
Specifications
Specifications
| Antigen | ZSCAN5 |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | PCRP-ZSCAN5A-2H4 |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495 |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa37-124) of human ZSCAN5 protein |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?