missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN5 Antibody (PCRP-ZSCAN5A-2H4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-27033CL1
This item is not returnable.
View return policy
Description
ZSCAN5 Monoclonal antibody specifically detects ZSCAN5 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry/Immunofluorescence
Specifications
| ZSCAN5 | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495 | |
| Recombinant fragment (around aa37-124) of human ZSCAN5 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| PCRP-ZSCAN5A-2H4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 79149 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction