missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-46728-25ul
This item is not returnable.
View return policy
Description
ZSCAN5 Polyclonal antibody specifically detects ZSCAN5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZSCAN5 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LEKDLEEDREENPGLTSPEPQLPNSPT | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79149 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction