missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN4 Antibody (OTI1A6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74957H
This item is not returnable.
View return policy
Description
ZSCAN4 Monoclonal specifically detects ZSCAN4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ZSCAN4 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ZSCAN4 | |
| Recombinant protein fragment corresponding to amino acids 164-433 of human ZSCAN4 (NP_689890) produced in E.coli. | |
| 0.1 mL | |
| 201516.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A6 | |
| Western Blot, Immunohistochemistry | |
| FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction