missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35200-20ul
This item is not returnable.
View return policy
Description
ZSCAN4 Polyclonal antibody specifically detects ZSCAN4 in Human samples. It is validated for ELISA,Western Blot
Specifications
| ZSCAN4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| FLJ35105, MGC126787, zinc finger and SCAN domain containing 4, Zinc finger protein 494MGC126789, ZNF494zinc finger and SCAN domain-containing protein 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-227 of human ZSCAN4 (NP_689890.1).,, Sequence:, MALDLRTIFQCEPSENNLGSENSAFQQSQGPAVQREEGISEFSRMVLNSFQDSNNSYARQELQRLYRIFHSWLQPEKHSKDEIISLLVLEQFMIGGHCNDKASVKEKWKSSGKNLERFIEDLTDDSINPPALVHVHMQGQEALFSEDMPLRDVIVHLTKQVNAQTTREANMGTPSQTSQDTSLETGQGYEDEQDGWNSSSKTTRVNENITNQGNQIVSLIIIQEENGPRPEEGGVSSDNPYNSKRAELVTARSQEGSINGITFQGVPMVMGAGCISQPEQSSPESALTHQSNEGNSTCEVHQKGSHGVQK | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 201516 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction