missing translation for 'onlineSavingsMsg'
Learn More

ZSCAN21/ZFP38 Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30734016 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734016 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734016 Supplier Novus Biologicals Supplier No. AF8015J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

ZSCAN21/ZFP38 Polyclonal antibody specifically detects ZSCAN21/ZFP38 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN21/ZFP38
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias DKFZp434L134, KOX25, NY-REN-21, Renal carcinoma antigen NY-REN-21, Zfp-38, ZFP38, zinc finger and SCAN domain containing 21, zinc finger and SCAN domain-containing protein 21, zinc finger protein 38, zinc finger protein 38 (KOX 25), Zinc finger protein 38 homolog, zinc finger protein NY-REN-21 antigen, Zipro1, ZNF38DKFZp686H10254
Host Species Sheep
Immunogen E. coli-derived recombinant human ZSCAN21, Asn139-Asp223, Accession # Q9Y5A6
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7589
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.