missing translation for 'onlineSavingsMsg'
Learn More

ZSCAN16 Antibody (4A9), Novus Biologicals™

Product Code. 18414229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414229 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414229 Supplier Novus Biologicals Supplier No. H00080345M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZSCAN16 Monoclonal antibody specifically detects ZSCAN16 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN16
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4A9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_079507
Gene Alias dJ265C24.3, FLJ22191, zinc finger and SCAN domain containing 16, zinc finger and SCAN domain-containing protein 16, Zinc finger protein 392ZNF392, Zinc finger protein 435ZNF435
Host Species Mouse
Immunogen ZNF435 (NP_079507.1, 132 a.a. ∽ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GNSERRDILMDKLAPLGRPYESLTVQLHPKKTQLEQEAGKPQRNGDKTRTKNEELFQKEDMPKDKEFLGEINDRLNKDTPQHPKSKDIIENEGRSEWQQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80345
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.