missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZSCAN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541696
This item is not returnable.
View return policy
Description
Peptide sequence: PECGKVFLHN SVLTEHGKIH LLEPPRKKAP RSKGPRESVP PRDGAQGPVA Sequence homology: Human: 100%.
ZSCAN1 may be involved in transcriptional regulation.
Specifications
| ZSCAN1 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN1 | |
| KOX29; MZF-1; zinc finger and SCAN domain containing 1; zinc finger and SCAN domain-containing protein 1; zinc finger with SCAN domain 1; ZNF31; ZNF360; ZNF915; ZSCAN1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 284312 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NBB4 | |
| ZSCAN1 | |
| Synthetic peptide directed towards the middle region of human ZSCAN1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction