missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZRSR2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57307
This item is not returnable.
View return policy
Description
ZRSR2 Polyclonal specifically detects ZRSR2 in Human samples. It is validated for Western Blot.
Specifications
| ZRSR2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CCCH type zinc finger, RNA-binding motif and serine/arginine rich protein 2, Renal carcinoma antigen NY-REN-20, U2 small nuclear ribonucleoprotein auxiliary factor 35 kDa subunit-relatedprotein 2, U2 small nuclear ribonucleoprotein auxiliary factor, small subunit 2, U2(RNU2) small nuclear RNA auxiliary factor 1-like 2MGC142014, U2AF1L2, U2AF1RS2MGC142040, U2AF1-RS2U2AF35-related protein, URPU2 small nuclear RNA auxiliary factor 1-like 2, zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2 | |
| Rabbit | |
| 52 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: -related protein 2. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Rabbit, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q15696 | |
| ZRSR2 | |
| Synthetic peptides corresponding to ZRSR2(zinc finger (CCCH type), RNA-binding motif and serine/arginine rich 2) The peptide sequence was selected from the N terminal of ZRSR2 (NP_005080). Peptide sequence LKEQWEEQQRKEREEEEQKRQEKKEKEEALQKMLDQAENELENGTTWQNP The peptide sequence for this immunogen was taken from within the described region. | |
| Affinity purified | |
| RUO | |
| 8233 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction