missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZRANB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55323
This item is not returnable.
View return policy
Description
ZRANB2 Polyclonal specifically detects ZRANB2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ZRANB2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Oocyte-specific maternal effect factor, zygote arrest 1, zygote arrest protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 9406 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation | |
| NP_005446.2 | |
| ZRANB2 | |
| The immunogen recognized by this antibody maps to a region between residue 270 and the C-terminus (residue 320) of human zinc-finger protein 265 (zinc-finger, splicing) using the numbering given in entry NP_005446.2 (GeneID 9406). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction