missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZRANB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598975
This item is not returnable.
View return policy
Description
ZRANB1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ZRANB1 is a positive regulator of the Wnt signaling pathway that specifically cleaves 'Lys-63'-linked ubiquitin chains. It acts by deubiquitinating APC protein, a negative regulator of Wnt-mediated transcription and may also modulate TNF-alpha signaling.
Specifications
| ZRANB1 | |
| Polyclonal | |
| Unconjugated | |
| ZRANB1 | |
| 9330160G10Rik; D7Wsu87e; DKFZp762P2216; Gm1956; hTrabid; Trabid; TRAF-binding domain-containing protein; TRAF-binding protein; TRAF-binding protein domain; ubiquitin thioesterase ZRANB1; Zinc finger Ran-binding domain-containing protein 1; zinc finger RANBP2-type containing 1; zinc finger, RAN-binding domain containing 1; ZRANB1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54764 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 3.75 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UGI0 | |
| ZRANB1 | |
| Recombinant Human Ubiquitin thioesterase ZRANB1 protein (111-397AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction