missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZPLD1 Monoclonal Antibody (1D8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524518
This item is not returnable.
View return policy
Description
Peptide Sequence: WNVLMDYCYT TPSGNPNDDI RYDLFLSCDK DPQTTVIENG RSQRGRFSF EVFRFVKHKN QKMSTVFLHCV TKLCRADDCP FLMPICSHRE RRDAGRRTTW.
ZPLD1 is a protein coding gene. Glycoprotein which is a component of the gelatinous extracellular matrix in the cupulae of the vestibular organ.
Specifications
| ZPLD1 | |
| Monoclonal | |
| Unconjugated | |
| ZPLD1 | |
| 9430016A21Rik; Cupulin; zona pellucida like domain containing 1; zona pellucida-like domain containing 1; zona pellucida-like domain-containing protein 1; Zona pellucida-like domain-containing protein 1, secreted form; ZP domain-containing protein 1; ZPLD1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 131368 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1D8 | |
| PBS with no preservative; pH 7.4 | |
| Q8TCW7 | |
| ZPLD1 | |
| ZPLD1 (NP_778226, 244 a.a. ~ 343 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction