missing translation for 'onlineSavingsMsg'
Learn More

ZPBP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-33671-25ul

 View more versions of this product

Product Code. 18407391

  • £328.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ZPBP2 Polyclonal specifically detects ZPBP2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZPBP2
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Q6X784
ZPBP2
This antibody was developed against a recombinant protein corresponding to amino acids: VELHQNSPVLICMDFKLSKKEIVDPTYLWIGPNEKTLTGNNRINITETGQLMVKDFLEPLSGLYTCTLSYKTVKAETQ
25 μL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
CAAX prenyl protease 1 homolog, EC 3.4.24.84, FACE1FLJ14968, FACE-1zinc metalloproteinase (STE24 homolog, yeast), Farnesylated proteins-converting enzyme 1, farnesylated-proteins converting enzyme 1, HGPS, Hutchinson-Gilford progeria syndrome, MADB, Prenyl protein-specific endoprotease 1, PRO1, Ste24p, STE24zinc metallopeptidase (STE24 homolog, yeast), zinc metallopeptidase (STE24 homolog, S. cerevisiae), Zinc metalloproteinase Ste24 homolog
Rabbit
Affinity Purified
RUO
124626
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only