missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZPBP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57768
This item is not returnable.
View return policy
Description
ZPBP2 Polyclonal specifically detects ZPBP2 in Human samples. It is validated for Western Blot.
Specifications
| ZPBP2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ZPBP2 | |
| Synthetic peptides corresponding to ZPBP2 (zona pellucida binding protein 2) The peptide sequence was selected from the middle region of ZPBP2)(50ug). Peptide sequence VRLDSCRPGFGKNERLHSNCASCCVVCSPATFSPDVNVTCQTCVSVLTYG. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Guinea pig: 92%; Equine: 92%; Mouse: 92%; Pig: 92%; Rat: 92%; Canine: 84%. | |
| Human, Mouse, Rat, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| CAAX prenyl protease 1 homolog, EC 3.4.24.84, FACE1FLJ14968, FACE-1zinc metalloproteinase (STE24 homolog, yeast), Farnesylated proteins-converting enzyme 1, farnesylated-proteins converting enzyme 1, HGPS, Hutchinson-Gilford progeria syndrome, MADB, Prenyl protein-specific endoprotease 1, PRO1, Ste24p, STE24zinc metallopeptidase (STE24 homolog, yeast), zinc metallopeptidase (STE24 homolog, S. cerevisiae), Zinc metalloproteinase Ste24 homolog | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 124626 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction