missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZP4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69341
This item is not returnable.
View return policy
Description
ZP4 Polyclonal specifically detects ZP4 in Human samples. It is validated for Western Blot.
Specifications
| ZP4 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| zinc finger and homeodomain protein 1, zinc fingers and homeobox 1, zinc fingers and homeoboxes 1, zinc fingers and homeoboxes protein 1, zinc-fingers and homeoboxes 1 | |
| Rabbit | |
| 49 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q12836 | |
| ZP4 | |
| Synthetic peptides corresponding to ZP4(zona pellucida glycoprotein 4) The peptide sequence was selected from the N terminal of ZP4. Peptide sequence MWLLRCVLLCVSLSLAVSGQHKPEAPDYSSVLHCGPWSFQFAVNLNQEAT. | |
| Affinity purified | |
| RUO | |
| 57829 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction