missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZP4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | ZP4 |
|---|---|
| Dilution | Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZP4 Polyclonal antibody specifically detects ZP4 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ZP4 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| zinc finger and homeodomain protein 1, zinc fingers and homeobox 1, zinc fingers and homeoboxes 1, zinc fingers and homeoboxes protein 1, zinc-fingers and homeoboxes 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human ZP4 (NP_067009.1). IPVQKALDLPFPSHHQRFSIFTFSFVNPTVEKQALRGPVHLHCSVSVCQPAETPSCVVTCPDLSRRRNFDNSSQNTTASVSSKGPMILLQATKDPPEKLRV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 57829 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title