missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZP3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15584642
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15584642 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15584642 Supplier Invitrogen™ Supplier No. PA541135

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: NKGDCGTPSH SRRQPHVMSQ WSRSASRNRR HVTEEADVTV GATDLPGQEW Sequence homology: Cow: 100%; Horse: 100%; Human: 100%; Rabbit: 100%; Rat: 90%.

The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in primary binding and induction of the sperm acrosome reaction. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a C-terminal consensus furin cleavage site, and a transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies. A variation in the last exon of this gene has previously served as the basis for an additional ZP3 locus; however, sequence and literature review reveals that there is only one full-length ZP3 locus in the human genome. Another locus encoding a bipartite transcript designated POMZP3 contains a duplication of the last four exons of ZP3, including the above described variation, and maps closely to this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZP3
Gene Accession No. P21754
Gene Alias Processed zona pellucida sperm-binding protein 3; Sperm receptor; zona pellucida C; zona pellucida C protein; zona pellucida glycoprotein 3; zona pellucida glycoprotein 3 (sperm receptor); zona pellucida glycoprotein 3 preproprotein; zona pellucida glycoprotein 3A (sperm receptor); zona pellucida glycoprotein 3B; zona pellucida glycoprotein ZP3; zona pellucida glycoprotein ZP3B; zona pellucida protein C; Zona pellucida sperm-binding protein 3; Zp3; Zp-3; ZP3A; ZP3A/ZP3B; ZP3B; Zp-3B; ZPC
Gene Symbols ZP3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZP3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7784
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.