missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £366.00
Specifications
| Antigen | ZP3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616991
|
Novus Biologicals
NBP2-94499-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689860
|
Novus Biologicals
NBP2-94499-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZP3 Polyclonal antibody specifically detects ZP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| ZP3 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| zona pellucida glycoprotein 3 (sperm receptor) | |
| A synthetic peptide corresponding to a sequence within amino acids 246-345 of human ZP3 (NP_001103824.1). DASSAFKVPRPGPDTLQFTVDVFHFANDSRNMIYITCHLKVTLAEQDPDELNKACSFSKPSNSWFPVEGSADICQCCNKGDCGTPSHSRRQPHVMSQWSR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 7784 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title