Learn More
Invitrogen™ ZP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587653
Description
Immunogen sequence: KMTVSLPGPI LLLSDDSSFR GVGSSDLKAS GSSGEKSRSE TGEEVGSRGA MDTKGHKTAG DVGSKAVAAV AAFAGVVATL GFIYYLYEKR TVSNH; Positive Samples: SH-SY5Y, HL-60, SW620, Mouse liver, Rat ovary; Cellular Location: Cell membrane, Single-pass type I membrane protein, extracellular matrix, extracellular space.
The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in secondary binding and penetration of acrosome-reacted spermatozoa. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a consensus furin cleavage site, and a C-terminal transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies.
Specifications
| ZP2 | |
| Polyclonal | |
| Unconjugated | |
| Zp2 | |
| 75-kDa rabbit zona pellucida protein; Processed zona pellucida sperm-binding protein 2; zona pellucida 2 glycoprotein; zona pellucida A protein; zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); zona pellucida glycoprotein ZP2; zona pellucida glycoprotein ZPA; zona pellucida protein A; Zona pellucida sperm-binding protein 2; ZP2; Zp-2; ZPA | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22787, 7783, 81828 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54767, P20239, Q05996 | |
| Zp2 | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.