missing translation for 'onlineSavingsMsg'
Learn More

ZP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15515652
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515652 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515652 Supplier Invitrogen Supplier No. PA542736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ZP2 Polyclonal Antibody

ZP2 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Zp2
Gene Accession No. Q05996
Gene Alias 75-kDa rabbit zona pellucida protein; Processed zona pellucida sperm-binding protein 2; zona pellucida 2 glycoprotein; zona pellucida A protein; zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); zona pellucida glycoprotein ZP2; zona pellucida glycoprotein ZPA; zona pellucida protein A; Zona pellucida sperm-binding protein 2; ZP2; Zp-2; ZPA
Gene Symbols Zp2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ZP2 (aa 553-602).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7783
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.