Learn More
Invitrogen™ ZP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595360
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat ovary tissue, SKOV3 whole cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The mammalian zona pellucida, which mediates species-specific sperm binding, induction of the acrosome reaction and prevents post-fertilization polyspermy, is composed of three to four glycoproteins, ZP1, ZP2, ZP3, and ZP4. ZP1 ensures the structural integrity of the zona pellucida.
Specifications
| ZP1 | |
| Polyclonal | |
| Unconjugated | |
| ZP1 | |
| epididymis luminal protein 163; HEL163; mgc87693; OOMD; OOMD1; Processed zona pellucida sperm-binding protein 1; Zona pellucida glycoprotein 1; zona pellucida glycoprotein 1 (sperm receptor); zona pellucida sperm-binding protein 1; ZP1; Zp-1; zp1 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22917, 85271 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide | |
| O54766, P60852 | |
| ZP1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human ZP1 (102-139aa HVLEKDGRFHLRVFMEAVLPNGRVDVAQDATLICPKPD). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.