missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536613
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.
The mammalian zona pellucida, which mediates species-specific sperm binding, induction of the acrosome reaction and prevents post-fertilization polyspermy, is composed of three to four glycoproteins, ZP1, ZP2, ZP3, and ZP4. ZP1 ensures the structural integrity of the zona pellucida.
Specifications
| ZP1 | |
| Polyclonal | |
| Unconjugated | |
| ZP1 | |
| epididymis luminal protein 163; HEL163; mgc87693; OOMD; OOMD1; Processed zona pellucida sperm-binding protein 1; Zona pellucida glycoprotein 1; zona pellucida glycoprotein 1 (sperm receptor); zona pellucida sperm-binding protein 1; ZP1; Zp-1; zp1 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22917 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P60852 | |
| ZP1 | |
| Synthetic peptide corresponding to amino acids 240-280 of human ZP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction