missing translation for 'onlineSavingsMsg'
Learn More

Zoxazolamine, MedChemExpress

Product Code. 16404587
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16404587 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16404587 Supplier Medchem Express Supplier No. HYB1307
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Zoxazolamine is widely used for a pharmacologic test that serves as a convenient indicator of changes in cytochrome P-450 activity in rodents.

Zoxazolamine is widely used for a pharmacologic test that serves as a convenient indicator of changes in cytochrome P-450 activity in rodents.

Specifications

Chemical Name or Material Zoxazolamine
CAS 61-80-3
Color White
Purity Grade Notes Research
Molecular Formula C7H5ClN2O
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (593.19 mM)
SMILES NC1=NC2=CC(Cl)=CC=C2O1
Molecular Weight (g/mol) 168.58
Formula Weight 168.58
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.