missing translation for 'onlineSavingsMsg'
Learn More

Zosuquidar Trihydrochloride, TRC

Product Code. 30456972
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456972 50 mg 50mg
30452099 10 mg 10mg
30452111 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456972 Supplier Toronto Research Chemicals Supplier No. TRCZ70087050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167465-36-3
Molecular Formula C32 H31 F2 N3 O2 . 3 Cl H
Molecular Weight (g/mol) 636.99
Synonym (αR)-4-[(1aα,6α,10bα)-1,1-Difluoro-1,1a,6,10b-tetrahydrodibenzo[a,e]cyclopropa[c]cyclohepten-6-yl]-α-[(5-quinolinyloxy)methyl]-1-piperazineethanol Hydrochloride, Zosuquidar Hydrochloride, LY 335979, RS 33295-198
SMILES Cl.Cl.Cl.O[C@@H](COc1cccc2ncccc12)CN3CCN(CC3)[C@H]4c5ccccc5[C@H]6[C@@H](c7ccccc47)C6(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C32H31F2N3O2.3ClH/c33-32(34)29-22-7-1-3-9-24(22)31(25-10-4-2-8-23(25)30(29)32)37-17-15-36(16-18-37)19-21(38)20-39-28-13-5-12-27-26(28)11-6-14-35-27;;;/h1-14,21,29-31,38H,15-20H2;3*1H/t21-,29-,30+,31-;;;/m1.../s1
Formula Weight 635.1685
Chemical Name or Material Zosuquidar trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.