missing translation for 'onlineSavingsMsg'
Learn More

Zorubicin Hydrochloride, TRC

Product Code. 30011206
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011206 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011206 Supplier Toronto Research Chemicals Supplier No. TRCZ70055010MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 36508-71-1
Molecular Formula C34 H35 N3 O10 . Cl H
Quantity 10 mg
Synonym Benzoic acid, 2-[1-[(2S,4S)-4-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-1,2,3,4,6,11-hexahydro-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-2-naphthacenyl]ethylidene]hydrazide, hydrochloride (1:1), Benzoic acid, [1-[(2S,4S)-4-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-1,2,3,4,6,11-hexahydro-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-2-naphthacenyl]ethylidene]hydrazide, monohydrochloride (9CI), Benzoic acid, [1-[4-[(3-amino-2,3,6-trideoxy-α-L-lyxo-hexopyranosyl)oxy]-1,2,3,4,6,11-hexahydro-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-2-naphthacenyl]ethylidene]hydrazide, monohydrochloride, (2S-cis)-, NSC 164011, RP 22050 hydrochloride, Rubidazone hydrochloride, Zorubicin hydrochloride
InChI Formula InChI=1S/C34H35N3O10.ClH/c1-15-28(38)20(35)12-23(46-15)47-22-14-34(44,16(2)36-37-33(43)17-8-5-4-6-9-17)13-19-25(22)32(42)27-26(30(19)40)29(39)18-10-7-11-21(45-3)24(18)31(27)41;/h4-11,15,20,22-23,28,38,40,42,44H,12-14,35H2,1-3H3,(H,37,43);1H/t15-,20-,22-,23-,28+,34-;/m0./s1
SMILES Cl.COc1cccc2C(=O)c3c(O)c4C[C@](O)(C[C@H](O[C@H]5C[C@H](N)[C@H](O)[C@H](C)O5)c4c(O)c3C(=O)c12)C(=NNC(=O)c6ccccc6)C
IUPAC Name N-[1-[(2S,4S)-4-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-2,5,12-trihydroxy-7-methoxy-6,11-dioxo-3,4-dihydro-1H-tetracen-2-yl]ethylideneamino]benzamide;hydrochloride
Molecular Weight (g/mol) 682.12
Formula Weight 681.2089
Chemical Name or Material Zorubicin Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.