missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide, TRC

Product Code. 30006682
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006682 50 mg 50mg
30008867 10 mg 10mg
30010807 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006682 Supplier Toronto Research Chemicals Supplier No. TRCZ70000050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68291-97-4
Formula Weight 212.0256
InChI Formula InChI=1S/C8H8N2O3S/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2,(H2,9,11,12)
IUPAC Name 1,2-benzoxazol-3-ylmethanesulfonamide
Molecular Formula C8 H8 N2 O3 S
Molecular Weight (g/mol) 212.23
Percent Purity >95%
SMILES NS(=O)(=O)Cc1noc2ccccc12
Synonym Zonisamide, 1,2-Benzisoxazole-3-methanesulfonamide, (1,2-Benzisoxazol-3-yl)methanesulfonamide, 3-(Sulfamoylmethyl)-1,2-benzisoxazole, AD 810, AD 810 (sulfonamide), Aleviatin, CI 912, Exceglan, Excegram, Excegran, PD 110843, Zonegran
Chemical Name or Material Zonisamide
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.