missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide

Product Code. 16514464 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16514464 250 mg 250mg
16504464 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16514464 Supplier Thermo Scientific Acros Supplier No. 465042500
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 68291-97-4 | C8H8N2O3S | 212.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68291-97-4
Molecular Formula C8H8N2O3S
Molecular Weight (g/mol) 212.22
MDL Number MFCD00865316
InChI Key UBQNRHZMVUUOMG-UHFFFAOYSA-N
IUPAC Name (1,2-benzoxazol-3-yl)methanesulfonamide
SMILES NS(=O)(=O)CC1=NOC2=CC=CC=C12

Specifications

Melting Point 165.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: .. Other solubilities: sparingly soluble in chloroform, n-hexane, soluble in methanol, ethanol, ethyl acetate and, acetic acid
Formula Weight 212.22
Chemical Name or Material Zonisamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.