missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide, Mikromol™

Product Code. 17980456
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17980456 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17980456 Supplier Mikromol Supplier No. MM1379.00
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Zonisamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zonisamide
CAS 68291-97-4
Analyte or Component Names Zonisamide
Shipping Condition Room Temperature
Molecular Formula C8 H8 N2 O3 S
Certifications/Compliance ISO 17025
Synonym Zonisamide, 1,2-Benzisoxazole-3-methanesulfonamide, (1,2-Benzisoxazol-3-yl)methanesulfonamide, 3-(Sulfamoylmethyl)-1,2-benzisoxazole, AD 810, AD 810 (sulfonamide), Aleviatin, CI 912, Exceglan, Excegram, Excegran, PD 110843, Zonegran
InChI Formula InChI=1S/C8H8N2O3S/c9-14(11,12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2,(H2,9,11,12)
SMILES NS(=O)(=O)Cc1noc2ccccc12
IUPAC Name 1,2-benzoxazol-3-ylmethanesulfonamide
Molecular Weight (g/mol) 212.23
Quantity 100 mg
Formula Weight 212.0256 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.